Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACADL Rabbit Polyclonal Antibody

Supplier: BioVision

3139-100
10003-442EA 262.65 USD InStock
10003-442
Anti-ACADL Rabbit Polyclonal Antibody
Antibodies
ACADL belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes. This protein is involved in fatty acid and branched chain amino acid metabolism. ACADL catalyzes the initial step of mitochondria beta-oxidation of straight-chain fatty acid.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ACADL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty