Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP9 Rabbit Polyclonal Antibody

Supplier: BioVision

3136-100
10003-430EA 262.65 USD InStock
10003-430
Anti-CASP9 Rabbit Polyclonal Antibody
Antibodies
Caspase family of cysteine proteases plays a key role in apoptosis. Caspase-9 is one of the most important caspases among the caspase family members. Upon induction of apoptosis, cytochrome c released from mitochondria associates with procaspase-9/Apaf-1. The complex processes procaspase-9 into a large subunit (35 kDa or 17-25 kDa) and small (10 kDa) by self-cleavage at D315. Activated caspase-9 further cleaves other caspase members including caspase-3, one of the proteases responsible for the proteolytic cleavage of many key proteins in apoptosis. In addition to self-cleavage, procaspase-9 can also be cleaved in vivo by caspase-3 at D330. The process served as a positive feedback to amplify the apoptotic signal in caspase activation pathway.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty