Anti-CASP7 Rabbit Polyclonal Antibody

Supplier: BioVision
3135-100
10003-426EA 286.04 USD
10003-426
Anti-CASP7 Rabbit Polyclonal Antibody
Caspase-7 is a 34 kDa protein consisting of two subunits and a short prodomain. The procaspase-7 can be activated by cleavage by upstream caspases to form active caspase-7 in apoptosis. Once activated, caspase-7 cleaves many of the same substrates as caspase-3, including poly (ADP-ribose) polymerase (PARP) and sterol regulatory element binding proteins (SREBPs).

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CASP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR