Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STAT1 Rabbit Polyclonal Antibody

Supplier: BioVision

3133R-100
10003-418EA 252.35 USD InStock
10003-418
Anti-STAT1 Rabbit Polyclonal Antibody
Antibodies
STATs (signal transducers and activators of transcription) are a family of cytoplamic latent transcription factors that are activated to regulate gene expression in response to a large number of extracellular signalling peptides including cytokines, interferons, and growth factors. After phosphorylation by JAK tyrosine kinases, STATs enter the nucleus to regulate transcription of many different genes. Among the seven STATs (Stat1, Stat2, Stat3, Stat4, Stat5a, Stat5b, and Stat6). Stat1, Stat3, Stat5a and Stat5b have wide activation profiles. Stat1 is activated by many different ligands including IFN family (IFN-a, IFN-b, IFN-r, and IL-10), gp130 family (IL-6, IL-11, LIF, CNTF, and G-CSF-1), and receptor tyrosine kinases (EGF, PDGF, and CSF-1). Stat1 has two forms, the 91 kDa STAT1α and the 84 kDa STAT1β, which are coded by the same gene with splicing variant.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty