Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DNA-PK Rabbit Polyclonal Antibody

Supplier: BioVision

3130-100
10003-416EA 272.95 USD InStock
10003-416
Anti-DNA-PK Rabbit Polyclonal Antibody
Antibodies
DNA-dependent protein kinase (DNA-PK) is made up of 3 subunits, 2 Ku proteins and a large catalytic subunit (DNA-PKcs). DNA-PKcs falls into the phosphatidyl-inositol 3-kinase superfamily althoμgh it does have serine/threonine protein kinase activity. This subunit migrates on SDS-PAGE gels as an intact and smaller processed form. In vitro, DNA-PK phosphorylates several transcription factors and other DNA-binding proteins and is believed to play a role in DNA damage recognition, repair and transcription. There is evidence to sμggest that DNA-PKcs may be a critical target for proteolysis by a caspase in apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DNA-PK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty