Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HNF4A Rabbit Polyclonal Antibody

Supplier: BioVision

3121-100
10003-398EA 334.75 USD InStock
10003-398
Anti-HNF4A Rabbit Polyclonal Antibody
Antibodies
HNF4A is a nuclear transcription factor which binds DNA as a homodimer. This protein controls the expression of several genes, including hepatocyte nuclear factor 1 alpha, a transcription factor which regulates the expression of several hepatic genes. This protein may play a role in development of the liver, kidney, and intestines. Mutations in the HNF4A gene have been associated with monogenic autosomal dominant non-insulin-dependent diabetes mellitus type I.

Recommended Dilutions: ELISA: 0.25 μg/ml; Western Blot: 2.5-5.0 μg/ml; Iimmunofluorescence: 1:10-1:50; Flow Cytometry: 1:10-1:50).

Type: Primary
Antigen: HNF4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty