Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Wee1 Rabbit Polyclonal Antibody

Supplier: BioVision

3120-100
10003-394EA 272.95 USD InStock
10003-394
Anti-Wee1 Rabbit Polyclonal Antibody
Antibodies
Wee1 is a nuclear tyrosine protein kinase. The protein plays a role in suppressing eukaryotes’ entry into mitosis by inactivating cdc2 kinase by inhibiting tyrosine phosphorylation. The predicted molecular weight of the kinase is 72 kDa. The kinase possesses tyrosine kinase activity, but structurally belongs to the serine/threonine protein kinase family.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in in Immunoprecipitation: 5-10 µg/ml

Type: Primary
Antigen: Wee1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty