Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RAF1 Rabbit Polyclonal Antibody

Supplier: BioVision

3116-100
10003-384EA 272.95 USD InStock
10003-384
Anti-RAF1 Rabbit Polyclonal Antibody
Antibodies
Human Raf1 is a proto-oncogene-encoded 74 kDa kinase that serves as an upstream activator of Mek1 by phosphorylating Ser-218 and Ser-222 in the MAP kinase kinase (Mek1). It has also been implicated in the phosphorylation and activation of the cdc2 tyrosine phosphatase cdc25. Raf1 binds to the GTP-bound form of the Ras G-protein via an N-terminal domain in Raf-1. Raf-1 is activated upon phosphorylation by protein kinase C, Src and ceramide-activated protein kinase. Raf-1 is highly related to the A-Raf and B-Raf isoforms within the N-terminal G-protein binding domain (CR1), phosphorylation site domain (CR2) and the catalytic domain (CR3). Raf-1 physically interacts with Ras, RapIB, MEK1, the 14-3-3 family of proteins and the molecular chaperones hsp90 and p50.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunoprecipitation: 15-25 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty