Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKA Rabbit Polyclonal Antibody

Supplier: BioVision

3115-100
10003-380EA 252.35 USD InStock
10003-380
Anti-PKA Rabbit Polyclonal Antibody
Antibodies
Protein kinase A (PKA) also known as cAMP-dependent protein kinase, is a 40 kDa protein kinase that phosphorylates serine or threonine residues in target proteins in response to elevated levels of cAMP. It has been sμggested that the complex structure of PKA may be necessary for it to accomplish such diverse functions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 20-30 µg/ml

Type: Primary
Antigen: PKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty