Anti-AMPK1 Rabbit Polyclonal Antibody

Supplier: BioVision
3113-100
10013-868EA 379.44 USD
10013-868
Anti-AMPK1 Rabbit Polyclonal Antibody
AMPKα (PRKAA1) belongs to the ser/thr protein kinase family. It is the catalytic subunit of the 5'-prime-AMP-activated protein kinase (AMPK). AMPK is a cellular energy sensor conserved in all eukaryotic cells. The kinase activity of AMPK is activated by the stimuli that increase the cellular AMP/ATP ratio. AMPK regulates the activities of a number of key metabolic enzymes throμgh phosphorylation. It protects cells from stresses that cause ATP depletion by switching off ATP-consuming biosynthetic pathways.

Recommended Dilutions: ELISA: 0.25 µg/ml; Western Blot: 2.5-5.0 µg/ml; Immunohistochemistry: 5 -25 µg/ml).

Type: Primary
Antigen: AMPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR