Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PYY3-36 Mouse Monoclonal Antibody [clone: RPY-B12]

Supplier: BioVision

3107-100
10003-372EA 272.95 USD InStock
10003-372
Anti-PYY3-36 Mouse Monoclonal Antibody [clone: RPY-B12]
Antibodies
Peptide YY (PYY), a member of a neuropeptide Y (NPY family) is a 36 AA peptide secreted from the L cells of the gastrointestinal tract in response to food intake. The release of PYY is stimulated by nutrients within the lower small intestine and the colon, which in turn decreases food intake by inhibiting gut motility. It acts as an “ileal brake” to cause a sense of satiety. PYY3-36 is produced by the dipeptidyl peptidase-IV digestion of polypeptide YY (PYY).

Recommended Dilutions: indirect ELISA for detection of antigen (recommended use at 1-2 µg/ml; Iimmunofluorescence: recommended use at 20-40µg/ml).

Type: Primary
Antigen: PYY3-36
Clonality: Monoclonal
Clone: RPY-B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty