Anti-Proinsulin Mouse Monoclonal Antibody [clone: HPI-B5]

3106-100
10003-370EA 288.4 USD
10003-370
Anti-Proinsulin Mouse Monoclonal Antibody [clone: HPI-B5]
Proinsulin is a prohormone precursor to Insulin, made by the pancreas in response to increased blood sμgar. Proinsulin is 39 amino acids longer than the mature Insulin, of which 4 are completely lost and the remaining 35 forms the C-peptide. The measurement of Proinsulin in serum provides a useful, valuable information for the diagnosis of insulinomas. Proinsulin levels have also been shown to be elevated in non-insulin dependent diabetics (NIDDM), in insulin dependent diabetics (IDDM) and other clinical situations. The antibody recognizes insulin and recombinant proinsulin, but does not react with synthetic C-peptide.

Recommended Dilutions: indirect ELISA, WB and IHC.

Type: Primary
Antigen: Proinsulin
Clonality: Monoclonal
Clone: HPI-B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR