Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IRS1 Mouse Monoclonal Antibody [clone: HIR-B1]

Supplier: BioVision

3105-100
10005-530EA 293.55 USD InStock
10005-530
Anti-IRS1 Mouse Monoclonal Antibody [clone: HIR-B1]
Antibodies
Insulin receptor substrates (IRS) subtypes are responsible for several insulin related activities, such as glucose homeostasis, cell growth, cell transformation, apoptosis and insulin signal transduction. The balance between Ser/Thr phosphorylation of IRS has been demonstrated to be an important regulator of insulin signaling. IRS1 has also been shown to be constitutively activated in cancers such as breast cancer, Wilm’s tumors, and adrenal cortical carcinomas. Does not recognize IRS peptide NOT phosphorylated at Ser616 and will not recognize IRS peptide phosphorylated at Tyr183. Also does not react with riboflavin carrier protein which is known to have multiple phosphate groups.

Recommended Dilutions: indirect ELISA and WB.

Type: Primary
Antigen: IRS1
Clonality: Monoclonal
Clone: HIR-B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty