Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-cGMP Rabbit Polyclonal Antibody

Supplier: BioVision

3568-100
10004-302EA 288.4 USD InStock
10004-302
Anti-cGMP Rabbit Polyclonal Antibody
Antibodies
Cyclic guanosine monophosphate (cGMP) serves as a second messenger in a manner similar to that observed with cAMP. Peptide hormones, such as the natriuretic factors, activate receptors that are associated with membrane-bound guanylate cyclase (GC). Receptor activation of GC leads to the conversion of GTP to cGMP. Nitric oxide (NO) also stimulates cGMP production by activating soluble GC, perhaps by binding to the heme moiety of the enzyme. Similar to cAMP, cGMP mediates most of its intracellular effects through the activation of specific cGMP dependent protein kinases (PKG).

Recommended Dilutions: 1:10-50 dilutions for nonradioactive EIA assays (10 µl/assay in an 70 µl assay reaction;The material is sufficient for performing 100-500 nonradioactive EIA assays or 6,000 radioactive assays. Detect 0.1-10 pmol/assay in a 70 µl assay reaction using nonradioactive method.

Type: Primary
Antigen: cGMP
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty