Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CAMP Rabbit Polyclonal Antibody

Supplier: BioVision

3567-100
10004-300EA 252.35 USD InStock
10004-300
Anti-CAMP Rabbit Polyclonal Antibody
Antibodies
Affinity purified rabbit IgG to 2’O-succinyl adenosine-3’,5’ cyclic monophosphate (cyclic AMP, cAMP).

  • Recommended Dilutions: 1:50 dilutions for nonradioactive EIA assays (10 µl/assay in an 70 µl assay reaction;The material is sufficient for performing 500 nonradioactive EIA assays or 6,000 radioactive assays. Detect 1-100 fmol/assay in a 70 µl assay reaction using nonradioactive method. Refer to the cAMP Assay Kit (K371-100) for detailed assay protocol.

Type: Primary
Antigen: CAMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty