Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TreakR Rabbit Polyclonal Antibody

Supplier: BioVision

3564-100
10004-294EA 272.95 USD InStock
10004-294
Anti-TreakR Rabbit Polyclonal Antibody
Antibodies
TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrates that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by biding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in aorta and liver. It may also play a role in vascular cell migration and angiogenesis.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: TreakR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty