Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF1A-associated Via Death Domain Rabbit Polyclonal Antibody

Supplier: BioVision

3561-100
10004-284EA 252.35 USD InStock
10004-284
Anti-TNFRSF1A-associated Via Death Domain Rabbit Polyclonal Antibody
Antibodies
TRADD (TNF-R1-associated death domain) is a 34 kDa protein that associates with the death domains of both FAS and TNF-R1 via their DDH regions. Overexpression of TRADD leads to NF?B activation and apoptosis in the absence of TNF.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TNFRSF1A-associated Via Death Domain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty