Anti-TLR9 Rabbit Polyclonal Antibody

Supplier: BioVision
3559R-100
10004-280EA 286.04 USD
10004-280
Anti-TLR9 Rabbit Polyclonal Antibody
The Toll-like receptor (TLR) family of proteins are characterized by a highly conserved Toll homology (TH) domain, which is essential for Toll-induced signal transduction. TLR9 has been sμggested as a receptor for unmethylated CpG dinucleotides in the bacterial DNA. TLR9 is highly expressed in the spleen.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TLR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR