Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR8 Mouse Monoclonal Antibody [clone: 44B05]

Supplier: BioVision

3558-100
10004-274EA 355.35 USD InStock
10004-274
Anti-TLR8 Mouse Monoclonal Antibody [clone: 44B05]
Antibodies
The Toll-like receptor (TLR) family of proteins are characterized by a highly conserved Toll homology (TH) domain, which is essential for Toll-induced signal transduction. TLR1 contains an extracellular domain consisting of several leucine-rich regions along with a single cytoplasmic Toll/IL-1R-like domain. TLR2 and TLR4 are activated in response to lipopolysacchride (LPS) stimulation, which results in the activation and translocation of NFkB and sμggests that these receptors are involved in mediating inflammatory responses. TLR6 is highly homologous to TLR1, and like other members of TLR family, it induces NFkB signaling upon activation. TLR8 gene contains three exons, two of which have coding function. Expression of TLR receptors is highest in peripheral blood leukocytes, macrophages, and monocytes.

Recommended Dilutions: Western Blot: 1:200-1000 dilutions

Type: Primary
Antigen: TLR8
Clonality: Monoclonal
Clone: 44B05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty