Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR2 Rabbit Polyclonal Antibody

Supplier: BioVision

3552R-100
10004-264EA 252.35 USD InStock
10004-264
Anti-TLR2 Rabbit Polyclonal Antibody
Antibodies
The Toll-like receptor (TLR) family in mammalian comprises a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and IL-1 receptor motif in the cytoplasmic domain. Up-to-date, ten TLRs (TLR1-10) have been described. TLR2 is differentially expressed in human cells. CD14+ monocytes expressed the highest level of TLR2 followed by CD15+ granulocytes, and CD19+ B-cells. CD3+ T-cells and CD56+ NK cells did not express TLR2. The expression of TLR2 on different cell types is regulated by different immune response modifiers. For example, LPS, GM-CSF, IL-1, and IL-10 up regulate TLR2, whereas IL-4, IFN-gamma, and TNF down regulate TLR2 expression in monocytes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TLR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty