Anti-Proteinase Inhibitor 9 Rabbit Polyclonal Antibody

Supplier: BioVision
3544R-100
10004-202EA 286.04 USD
10004-202
Anti-Proteinase Inhibitor 9 Rabbit Polyclonal Antibody
Proteinase inhibitor 9 (PI-9, also designated cytoplasmic antiproteinase 3, or CAP3) is a 42 kDa member of the ovalbumin family of serpins that is expressed in placenta, lung and cytotoxic lymphocytes. PI-9 is a potent inhibitor of granzyme B and of granzyme B-mediated apoptosis, and is also an inhibitor of capase-1 and, to a smaller extent, caspase-4 and caspase-8. PI-9 expression is also upregulated in response to inflammatory stimuli. This upregulation protects cells from apoptosis induced by endogenously expressed or released granzyme B, particulary during target cell killing

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: Proteinase Inhibitor 9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR