Anti-Proteinase Inhibitor 9 Mouse Monoclonal Antibody [clone: PI9-17]

Supplier: BioVision
3544-100
10004-200EA 343.88 USD
10004-200
Anti-Proteinase Inhibitor 9 Mouse Monoclonal Antibody [clone: PI9-17]
Proteinase inhibitor 9 (PI-9, also designated cytoplasmic antiproteinase 3, or CAP3) is a 42 kDa member of the ovalbumin family of serpins that is expressed in placenta, lung and cytotoxic lymphocytes. PI-9 is a potent inhibitor of granzyme B and of granzyme B-mediated apoptosis, and is also an inhibitor of capase-1 and, to a smaller extent, caspase-4 and caspase-8. PI-9 expression is also upregulated in response to inflammatory stimuli. This upregulation protects cells from apoptosis induced by endogenously expressed or released granzyme B, particulary during target cell killling.

Recommended Dilutions: Western Blot: 2-4 µg/ml; Immunohistochemistry: 20 µg/ml

Type: Primary
Antigen: Proteinase Inhibitor 9
Clonality: Monoclonal
Clone: PI9-17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR