Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK3 Rabbit Polyclonal Antibody

Supplier: BioVision

3542-100
10004-196EA 272.95 USD InStock
10004-196
Anti-MAPK3 Rabbit Polyclonal Antibody
Antibodies
Both p44 and p42 MAP kinases (Erk1 and Erk2) function in a protein kinase cascade that plays a critical role in the regulation of cell growth and differentiation. Activation of MAP kinases occurs throμgh phosphorylation of threonine and tyrosine at the sequence T*EY* by a single upstream MAP kinase kinase (MEK).

Recommended Dilutions: Western Blot: 1 µg/ml; Immunoprecipitation: 10-20 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty