Anti-NSF Rabbit Polyclonal Antibody

Supplier: BioVision
3541-100
10004-192EA 286.04 USD
10004-192
Anti-NSF Rabbit Polyclonal Antibody
Syntaxins were originally thoμght to be docking proteins, but have more recently been categorized as anchoring proteins that anchor themselves to the cytoplasmic surfaces of cellular membranes. Syntaxins have been shown to bind to various proteins involved in exocytosis, including VAMPs (vesicle-associated membrane proteins), NSF (N-ethylmaleimidesensitive factor), SNAP 25 (synaptosomal-associated protein of 25kDa), SNAPs (soluble NSF attachment proteins) and synaptotagmin. SNAPs mediate the membrane binding of NSF, which is essential for membrane fusion reactions. An additional protein designated synaptophysin may regulate exocytosis by competing with SNAP 25 and syntaxins for VAMP binding.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 5-15 mg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: NSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR