Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACADS Rabbit Polyclonal Antibody

Supplier: BioVision

3539-100
10004-188EA 252.35 USD InStock
10004-188
Anti-ACADS Rabbit Polyclonal Antibody
Antibodies
ACADS (acyl-Coenzyme A dehydrogenase), is a homotetrameric mitochondrial flavoprotein that belongs to the acyl-CoA dehydrogenase family. This enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Mutation of this gene causes SCAD deficiency an acute acidosis and muscle weakness in infants and lipid-storage myopathy in adults.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ACADS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty