Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACAA2 Rabbit Polyclonal Antibody

Supplier: BioVision

3538-100
10004-184EA 252.35 USD InStock
10004-184
Anti-ACAA2 Rabbit Polyclonal Antibody
Antibodies
ACAA2 (acetylcoenzyme A acyltransferase 2), is member of the thiolase family of enzymes and is involved in lipid metabolism. ACAA2 is localized in the mitochondria. ACCA2 catalyzes the last conversion step of acetyl-CoA to 3-oxoacyl-CoA, in the fatty acid oxidation pathway.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ACAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty