Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP8 Rabbit Polyclonal Antibody

Supplier: BioVision

3528-100
10004-170EA 272.95 USD InStock
10004-170
Anti-MMP8 Rabbit Polyclonal Antibody
Antibodies
The matrix metalloproteinases (MMP) are a family of peptidase enzymes responsible for the degradation of extracellular matrix components, including collagen, gelatin, fibronectin, laminin and proteoglycan. Transcription of MMP genes is differentially activated by phorbol ester, lipopolysaccharide (LPS) or staphylococcal enterotoxin B (SEB). MMP catalysis requires both calcium and zinc. Unlike other members of the MMP family, MMP-8 is expressed exclusively in inflammatory conditions. MMP-8 is also the predominant collagenase expressed in ulcers and healing wounds.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 15-25 µg/ml

Type: Primary
Antigen: MMP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty