Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP3 Mouse Monoclonal Antibody [clone: C33]

Supplier: BioVision

3004-100
10003-200EA 347.21 USD InStock
10003-200
Anti-CASP3 Mouse Monoclonal Antibody [clone: C33]
Antibodies
The caspase family of cysteine proteases play a key role in apoptosis. Caspase-3 is the most extensively studied apoptotic protein among caspase family members. Caspase-3 is synthesized as inactive pro-enzyme that is processed in cells undergoing apoptosis by self-proteolysis and/or cleavage by another upstream protease. The processed form of caspase-3 consists of large (17 kD) and small (12 kD) subunits which associate to form an active enzyme. The active caspase-3 proteolytically cleaves and activates other caspases, as well as relevant targets in the cells (e.g., PARP and DFF).

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CASP3
Clonality: Monoclonal
Clone: C33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty