Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PARP1 Mouse Monoclonal Antibody [clone: 10H]

Supplier: BioVision

3000-100
10003-194EA 303.85 USD InStock
10003-194
Anti-PARP1 Mouse Monoclonal Antibody [clone: 10H]
Antibodies
PARP, a 116 kDa nuclear poly (ADP-ribose) polymerase, is a highly conserved nuclear enzyme implicated in DNA repair and in the apoptosis response of cells. This protein can be cleaved by many caspases in vitro and is one of the main cleavage targets of caspase-3 in vivo. The cleavage occurs between ASP214 and Gly 215, which separates PARP’s N-terminal DNA binding domain (24 kDa) from its C-terminal catalytic domain (89 kDa). It has been shown that cleavage of PARP facilitates cellular disassembly and inhibition of PARP cleavage attenuates apoptosis in vitro.

Recommended Dilutions: Immunocytochemistry : 5-20 μg/ml; Western Blot: 10 μg/ml using colorimetric methods

Type: Primary
Antigen: PARP1
Clonality: Monoclonal
Clone: 10H
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty