Anti-BCL2 Mouse Monoclonal Antibody [clone: Bcl-2/100]

Supplier: BioVision
3195-100
10003-532EA 332.74 USD
10003-532
Anti-BCL2 Mouse Monoclonal Antibody [clone: Bcl-2/100]
Bcl-2 (B-cell leukemia/lymphoma 2) gene was discovered by walking the chromosomal breakpoints t(14;18). This gene consists of two exons and several transcripts that encode two different protein products, Bcl-2α and Bcl-2β. Both proteins are approximately 26-30 kDa and differ at their C-terminal region. Bcl-2 suppresses apoptosis (programmed cell death) and this property appears to be modulated by homodimerization or association with Bax and Bad, two cell death promoters.

Recommended Dilutions: Western Blot: 1-4 µg/ml;

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: Bcl-2/100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR