Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRKA Rabbit Polyclonal Antibody

Supplier: BioVision

3194-100
10003-528EA 252.35 USD InStock
10003-528
Anti-TRKA Rabbit Polyclonal Antibody
Antibodies
TrkA, the high affinity Nerve Growth Factor (NGF) receptor, autophosphorylates on tyrosine to activate multiple effectors. Phosphorylation at Tyr490 is required for Shc association and activation of the Ras-MAP kinase cascade. Residues Tyr674/675 lie within the catalytic domain, and phosphorylation at these sites reflects Trk kinase activity.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TRKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty