Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP3K14 Rabbit Polyclonal Antibody

Supplier: BioVision

3193R-100
10003-524EA 262.65 USD InStock
10003-524
Anti-MAP3K14 Rabbit Polyclonal Antibody
Antibodies
NIK (NF-kB-inducing kinase) is a member of the MAP kinase kinase kinase family that binds TRAF2 and stimulates NF-kB activity. NIK was initially isolated from a human B cell cDNA library and contains 795 amino acids with an apparent molecular weight of slightly more than 97 kDa on SDS gel. NIK is a serine/threonine kinase and its kinase activity contributes to IkB phosphorylation. The carboxyl terminal segment of NIK binds TRAF2. A mutant NIK with intact carboxyl terminus but without the two lysine residues at its catalytic domain serves as a dominant-negative inhibitor for NF-kB activation. NIK also interacts with TRAF6 and mediates IL-1-induced NF-kB activation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MAP3K14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Hamster

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty