Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTSL Rabbit Polyclonal Antibody

Supplier: BioVision

3192-100
10003-520EA 262.65 USD InStock
10003-520
Anti-CTSL Rabbit Polyclonal Antibody
Antibodies
Cathepsin L is a normal lysosomal cysteine endoproteases that is expressed in all cells. The latent 39 kDa pro-Cathepsin L must be processed by cleavage of the 96 amino acid pro-peptide, to generate the active two-chain form made up of 21 and 5 kDa subunits. Mature Cathepsin L is normally localized in the lysosomes where it plays a major role in intracellular protein catabolism. The fact that Cathepsin L is also capable of degrading extracellular matrix proteins sμggests that it may play a role in invasion and metastasis and could be a useful prognostic indicator for cancer.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; and Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: CTSL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty