Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTSD Rabbit Polyclonal Antibody

Supplier: BioVision

3191R-100
10003-516EA 262.65 USD InStock
10003-516
Anti-CTSD Rabbit Polyclonal Antibody
Antibodies
Cathepsin D is a normal lysosomal protease that is expressed in all cells. It is an aspartyl protease with a pH optimum in the range of 3-5, and contains two N-linked oligosaccharides. Cathepsin D is synthesized as an inactive 52 kDa pro-enzyme. Activation involves the proteolytic removal of the 43 amino acid profragment and an internal cleavage to generate the two-chain form made up of 34 and 14 kDa subunits. Cathepsin D contains the mannose-6-phosphate lysosomal localization signal that targets the enzyme to the lysosomal compartment where it functions in the normal degradation of proteins. In certain tumor cells, Cathepsin D is abnormally processed and is secreted in its 52 kDa precursor form. Numerous clinical studies as well as in vitro evidence sμggest that cathepsin D plays an important role in malignant transformation and may be a useful prognostic indicator for breast cancer.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in immunoprecipitation

Type: Primary
Antigen: CTSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty