Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTSB Rabbit Polyclonal Antibody

Supplier: BioVision

3190-100
10003-512EA 272.95 USD InStock
10003-512
Anti-CTSB Rabbit Polyclonal Antibody
Antibodies
Cathepsin B is a normal lysosomal cysteine proteinase that is expressed in all cells. It is synthesized as an inactive 43 kDa pro-enzyme which, by removal of a 62 amino acid propeptide, is activated to the single-chain form of 31 kDa or the two-chain form (25 and 5 kDa subunits). Matured cathepsin B is normally localized in the lysosomes where it functions in protein turnover. Cathepsin B is also shown capable of degrading extracellular matrix proteins. Elevated levels of cathepsin B correlate with malignancy sμggesting that this enzyme may be a useful prognostic marker for several types of human cancers.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml) amd in Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: CTSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty