Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRF1 Rabbit Polyclonal Antibody

Supplier: BioVision

3188R-100
10003-504EA 252.35 USD InStock
10003-504
Anti-PRF1 Rabbit Polyclonal Antibody
Antibodies
Human perforin (cytolysin) is believed to be one of the major effector molecules used by cytotoxic T cells and NK cells to mediate targeted cell lysis. Perforin expression is constitutive on NK cells, but increases in resting XD8+ cytotoxic cells upon activation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty