Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRF1 Mouse Monoclonal Antibody [clone: ?G9]

Supplier: BioVision

3188-100
10003-502EA 303.85 USD InStock
10003-502
Anti-PRF1 Mouse Monoclonal Antibody [clone: ?G9]
Antibodies
Human perforin (cytolysin) is believed to be one of the major effector molecules used by cytotoxic T cells and NK cells to mediate targeted cell lysis. Perforin expression is constitutive on NK cells, but increases in resting XD8+ cytotoxic cells upon activation.

Recommended Dilutions: Immunoprecipitation: 2-8 µg/ml; Flow Cytometry: 2-8 µg/ml; Immunohistochemistry: 2-8 µg/ml for both frozen and paraffin sections; Immunocytochemistry: 2-8 µg/ml

Type: Primary
Antigen: PRF1
Clonality: Monoclonal
Clone: ?G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty