Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HCCS Mouse Monoclonal Antibody [clone: N/A]

Supplier: BioVision

3187-100
10003-500EA 334.75 USD InStock
10003-500
Anti-HCCS Mouse Monoclonal Antibody [clone: N/A]
Antibodies
The precursor of cytochrome c, apocytochrome c, is synthesized in the cytoplasm. Upon translocation into mitochondria, cytochrome c acquires a heme moiety required for functionality in the mitochondrial respiration chain. The heme-bound form of cytochrome c is called holocytochrome c. When released into the cytosol, cytochrome c can activate caspases responsible for apoptosis.

Recommended Dilutions: Western Blot: 5-10 µg/ml;

Type: Primary
Antigen: Holocytochrome c
Clonality: Monoclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty