Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-XIAP Rabbit Polyclonal Antibody

Supplier: BioVision

3183-100
10003-494EA 262.65 USD InStock
10003-494
Anti-XIAP Rabbit Polyclonal Antibody
Antibodies
BIRC-4 (for baculoviral IAP repeat-containing protein 4) is a member of the IAP (for Inhibitor of apoptosis protein) family proteins. Other Aliases include RP1-315G1.5, API3, ILP1, MIHA, XIAP, XLP2. BIRC4 inhibits activated caspase-3 and apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in immunoprecipitation and Immunohistochemistry

Type: Primary
Antigen: XIAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty