Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-cIAP-2/HIAP-1 Rabbit Polyclonal Antibody

Supplier: BioVision

3181-100
10003-490EA 252.35 USD InStock
10003-490
Anti-cIAP-2/HIAP-1 Rabbit Polyclonal Antibody
Antibodies
Apoptosis can be inhibited by a group of proteins called inhibitors of apoptosis (IAPs). These proteins contain a BIR (baculovirus IAP repeat) domain near the amino-terminus. The BIR domain can bind some caspases. Many members of the IAP family of proteins block proteolytic activation of caspase-3 and –7. For example, XIAP, cIAP-1 and cIAP-2 appear to block cytochrome c-induced activation of caspase-9, thereby preventing initiation of the caspase cascade. Since cIAP-1 and cIAP-2 were first identified as components in the cytosolic death domain-induced complex associated with the TNF family of receptors, they may inhibit apoptosis by additional mechanisms.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: cIAP-2/HIAP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty