Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40L Rabbit Polyclonal Antibody

Supplier: BioVision

3178-100
10005-570EA 288.4 USD InStock
10005-570
Anti-CD40L Rabbit Polyclonal Antibody
Antibodies
CD40, also known as bp50 or CDW40, is a type I membrane protein found on the surface of B cells and primary carcinomas. It belongs to the TNF-R family. CD40 serves as the receptor for CD40 ligand (CD40L), a cytokine expressed on the cell surface of T cells. CD40 plays a critical role in B cell proliferation, antibody class switching, modulation of apoptosis in the germinal center throμgh interaction with T cells expressing CD40L, and activation of CD4+ T cells. Ligation of CD40 with its ligand preferentially induces stress-activated protein kinases in B lymphocytes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CD40L
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty