Anti-BLK Rabbit Polyclonal Antibody

Supplier: BioVision
3175-100
10005-562EA 332.74 USD
10005-562
Anti-BLK Rabbit Polyclonal Antibody
BLK, a BH3-containing protein that interacts with Bcl-2 and Bcl-xL, is a potent death agonist.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BLK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR