Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BID Rabbit Polyclonal Antibody

Supplier: BioVision

3172-100
10005-556EA 252.35 USD InStock
10005-556
Anti-BID Rabbit Polyclonal Antibody
Antibodies
Bid, a BH3 domain-containing proapoptotic Bcl-2 family member, is localized in the cytosolic fraction of cells as an inactive precursor. Its active form is generated upon proteolytic cleavage by caspase-8 in the Fas signaling pathway. Cleaved Bid translocates to mitochondria and releases its potent proapoptotic activity, which in turn induces cytochrome c release and mitochondrial damage. The cytochrome c releasing activity of Bid was antagonized by Bcl-2. Mutation in the SH3 domain can diminish the cytochrome c releasing activity. In the animal model studies, Bid-deficient mice are found resistant to the lethal effects of death factor signals relayed throμgh Fas.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 5-10 µg/ml; and Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: BID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty