Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BIRC5 Rabbit Polyclonal Antibody

Supplier: BioVision

3170-100
10005-552EA 252.35 USD InStock
10005-552
Anti-BIRC5 Rabbit Polyclonal Antibody
Antibodies
Survivin is a newly described apoptosis inhibitor that is expressed in many human cancers, but undetectable in terminally differentiated adult tissues. It has been shown that recombinant expression of Survivin counteracts apoptosis of B lymphocyte precursors deprived of interleukin 3 (IL-3), sμggesting a potential role of Survivin in cancer therapy. Survivin is expressed in G2-M phase in a cell cycle-dependent manner and directly associated with mitotic spindle microtubules, sμggesting a role in both apoptosis regulation and cell cycle progression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty