Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NANOG Rabbit Polyclonal Antibody

Supplier: BioVision

3165-100
10005-546EA 334.75 USD InStock
10005-546
Anti-NANOG Rabbit Polyclonal Antibody
Antibodies
NANOG is a transcription regulator involved with inner cell mass and embryonic stem (ES) cell proliferation and self-renewal. It imposes pluripotency on ES cells and prevents their differentiation towards extra-embryonic endoderm and trophectoderm lineages. This protein blocks bone morphogenetic protein-induced mesoderm differentiation of ES cells by physically interacting with SMAD1 and interfering with the recruitment of coactivators to the active SMAD transcriptional complexes.

Recommended Dilutions: ELISA: 0.25 µg/ml; Western Blot: 2.5-5.0 µg/ml).

Type: Primary
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty