Anti-BIRC5 Rabbit Polyclonal Antibody

Supplier: BioVision
3161-100
10005-538EA 309.39 USD
10005-538
Anti-BIRC5 Rabbit Polyclonal Antibody
Survivin is a newly described apoptosis inhibitor that is expressed in many human cancers, but undetectable in terminally differentiated adult tissues. It has been shown that recombinant expression of Survivin conteracts apoptosis of B lymphocyte precursors deprived of interleukin 3 (IL-3), sμggesting a potential role of Survivin in cancer therapy. Survivin interacts with the processed form of caspase-3 and inhibits its proteolytic activity.

Recommended Dilutions: Western Blot: 0.5 - 2 µg/ml

Type: Primary
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR