Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACADSB Rabbit Polyclonal Antibody

Supplier: BioVision

3157-100
10003-476EA 252.35 USD InStock
10003-476
Anti-ACADSB Rabbit Polyclonal Antibody
Antibodies
Short/branched chain acyl-CoA dehydrogenase(ACADSB) belongs to the acyl-CoA dehydrogenase family of enzymes catalyze the dehydrogenation of acyl-CoA derivatives in the metabolism of fatty acids or branch chained amino acids. ACADSB catalyzes the degradation of L-isoleucine and has the greatest affinity towards (S)-2-methylbutyryl-CoA, isobutyryl-CoA and 2-methylhexanoyl-CoA as substrates.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: ACADSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty