Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP6 Rabbit Polyclonal Antibody

Supplier: BioVision

3156-100
10003-474EA 390.6 USD InStock
10003-474
Anti-CASP6 Rabbit Polyclonal Antibody
Antibodies
Caspase family of cysteine proteases has been shown to play a key role in apoptosis. Similar to other caspases, caspase-6 is also synthesized as an inactive pro-enzyme that is processed in cells undergoing apoptosis. Together with caspase-3, caspase-6 is one of the major caspases in apoptotic cells, and functions downstream of apoptosis inhibitors Bcl-2 and Bcl-xL. Caspase-6 has also been shown involving in the proteolysis of poly (ADP-ribose) polymerase (PARP) and nuclear lamin A.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunofluorescence

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty