Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NHERF1/ EBP50 Rabbit Polyclonal Antibody

Supplier: BioVision

3521-100
10004-100EA 252.35 USD InStock
10004-100
Anti-NHERF1/ EBP50 Rabbit Polyclonal Antibody
Antibodies
NHE3 regulatory factor 1 (NHERF-1) is a multifunctional adaptor protein that interacts with NHE3 via its PDZ domains. The amino-terminal PDZ domain regulates the dimerization of NHERF-1 in vivo. G protein-coupled receptor kinase 6A phosphorylates NHERF-1 at Ser 289 via a PDZ domain-mediated interaction.

Recommended Dilutions: Western Blot: 1-4 µg/ml;

Type: Primary
Antigen: NHERF1/ EBP50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty