Anti-MAP2K1 + MAP2K2 Rabbit Polyclonal Antibody

Supplier: BioVision
3519-100
10004-098EA 402.79 USD
10004-098
Anti-MAP2K1 + MAP2K2 Rabbit Polyclonal Antibody
MAP kinase kinase 1 (Mek1), also known as mitogen activated protein kinase kinase, MAP kinase kinase, ERK kinase 1 or microtubule-associated protein 2 kinase, is a 43 kDa dual-specificity protein kinase that phosphorylates threonine and tyrosine residues on MAP kinases such as Erk 1 and 2. Mek1 and Mek2 are involved in a diverse array of cellular processes such as stress-activated response, apoptosis, cytokine-induced cell proliferation, and DNA recombination during meiosis. Mek1 and Mek2 are activated by phosphorylation on serine 218 and 222 by serine-threonine kinase, Raf-1, which is part of the p21ras signal transduction pathway.

Recommended Dilutions: Western Blot: 1:200-1000; Immunocytochemistry: 1:100-200

Type: Primary
Antigen: MAP2K1 + MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR